Learn More
Grainger G3261 Level Gauge HDPE D 79in
Catalog No. 502764501
Encompass_Preferred
The Grainger 16N344 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron frame, end brackets and conduit box. The 16N344 operates on single phase 115V power and has a full load current rating of 9.8 Amps. This electric motor is ideal for driving pumps, fans, compressors, conveyors and other industrial equipment.
Catalog No. 50-276-4501 Supplier Grainger Supplier No. 16N344
The Grainger 16N344 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is thermally protected and rated for continuous duty applications. It features a cast iron frame, end brackets and conduit box. The 16N344 operates on single phase 115V power and has a full load current rating of 9.8 Amps. This electric motor is ideal for driving pumps, fans, compressors, conveyors and other industrial equipment.