Learn More
Grainger G3261 Level Gauge HDPE D 82in
Catalog No. 502763918
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without opening the lid. Models include polypropylene and stainless steel construction with clear polycarbonate or acrylic sight tubes. Gauges feature 1" NPT connections and are available in 6" to 72" lengths to fit small to large capacity drums and tanks. They can handle temperatures up to 200°F and pressures up to 100 PSI. Overall, these gauges provide a safe, convenient way to monitor liquid levels in industrial containers.
Catalog No. 50-276-3918 Supplier Grainger Supplier No. 16N347
The Grainger website features liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without opening the lid. Models include polypropylene and stainless steel construction with clear polycarbonate or acrylic sight tubes. Gauges feature 1" NPT connections and are available in 6" to 72" lengths to fit small to large capacity drums and tanks. They can handle temperatures up to 200°F and pressures up to 100 PSI. Overall, these gauges provide a safe, convenient way to monitor liquid levels in industrial containers.