Learn More
Grainger G3261 Level Gauge HDPE D 88in
Catalog No. 502764683
Encompass_Preferred
The Grainger G3261 Level Gauge is a durable HDPE dipstick designed for measuring liquid levels in tanks and reservoirs. It features large, easy-to-read markings and has a total length of 88 inches. The gauge has a D profile that prevents it from rolling and allows it to sit upright. It is made from high density polyethylene plastic which provides chemical resistance. The gauge has black markings in both inches and centimeters for convenient reading in US and metric units. With its sturdy construction and clear markings, this gauge is ideal for monitoring fluid levels in a variety of industrial and commercial applications.
Catalog No. 50-276-4683 Supplier Grainger Supplier No. 16N352
The Grainger G3261 Level Gauge is a durable HDPE dipstick designed for measuring liquid levels in tanks and reservoirs. It features large, easy-to-read markings and has a total length of 88 inches. The gauge has a D profile that prevents it from rolling and allows it to sit upright. It is made from high density polyethylene plastic which provides chemical resistance. The gauge has black markings in both inches and centimeters for convenient reading in US and metric units. With its sturdy construction and clear markings, this gauge is ideal for monitoring fluid levels in a variety of industrial and commercial applications.