Learn More
Grainger G3261 Level Gauge HDPE D 91in
Catalog No. 502763923
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These affordable gauges install easily and are available in different sizes to fit standard drums and tanks. Overall, they are simple, durable liquid level indicators ideal for industrial applications.
Catalog No. 50-276-3923 Supplier Grainger Supplier No. 16N355
Add to Cart
Add to Cart
The Grainger website features liquid level gauges for drums and tanks. These gauges are made of galvanized steel or HDPE and use a rod or float mechanism to indicate the fluid level inside a container. Key features include compatibility with various liquids like oil, solvents and water. They resist corrosion and provide accurate, reliable level readings. These affordable gauges install easily and are available in different sizes to fit standard drums and tanks. Overall, they are simple, durable liquid level indicators ideal for industrial applications.