Learn More
Grainger G3261 Level Gauge HDPE D 92in
Catalog No. 502764135
Encompass_Preferred
The Grainger website features liquid level gauges for drums and tanks made of galvanized steel and HDPE. These rod and float gauges allow visual monitoring of liquid levels. They feature corrosion-resistant materials suitable for use with water, chemicals, petroleum products and more. Lengths range from 7" to 78" to fit different size containers. They have standard NPT thread connections and some feature flanged ends. Overall, these are basic mechanical float gauges for simple liquid level indication in drums and tanks.
Catalog No. 50-276-4135 Supplier Grainger Supplier No. 16N356
The Grainger website features liquid level gauges for drums and tanks made of galvanized steel and HDPE. These rod and float gauges allow visual monitoring of liquid levels. They feature corrosion-resistant materials suitable for use with water, chemicals, petroleum products and more. Lengths range from 7" to 78" to fit different size containers. They have standard NPT thread connections and some feature flanged ends. Overall, these are basic mechanical float gauges for simple liquid level indication in drums and tanks.