Learn More
Grainger G3261 Level Gauge HDPE D 93in
Catalog No. 502764684
Encompass_Preferred
The Grainger website offers a variety of liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without having to open the container. Options include polypropylene and stainless steel construction, clear and frosted acrylic lens options, gauges with or without valves, and gauges designed for specific liquid types like chemicals or petroleum. Gauges feature direct mount or flange mount installation. Lengths range from 7" to 48". These gauges are useful for monitoring liquid levels in industrial drums and tanks.
Catalog No. 50-276-4684 Supplier Grainger Supplier No. 16N357
The Grainger website offers a variety of liquid level gauges for drums and tanks. These gauges allow users to visually check liquid levels in their containers without having to open the container. Options include polypropylene and stainless steel construction, clear and frosted acrylic lens options, gauges with or without valves, and gauges designed for specific liquid types like chemicals or petroleum. Gauges feature direct mount or flange mount installation. Lengths range from 7" to 48". These gauges are useful for monitoring liquid levels in industrial drums and tanks.