Learn More
Grainger G3261 Level Gauge HDPE D 97in
Catalog No. 502764023
Encompass_Preferred
The Krueger Level Gauge For 97 in Container is a float-operated mechanical level gauge designed for use with 97 inch high containers. It features a hermetically sealed reed switch that activates indicator lights to provide a visual display of liquid level. This stainless steel gauge has an adjustable stem length from 34.5 to 48 inches and provides high, mid and low level indication. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger Level Gauge is ideal for use with hydraulic fluids, diesel fuel and motor oils.
Catalog No. 50-276-4023 Supplier Grainger Supplier No. 16N360
The Krueger Level Gauge For 97 in Container is a float-operated mechanical level gauge designed for use with 97 inch high containers. It features a hermetically sealed reed switch that activates indicator lights to provide a visual display of liquid level. This stainless steel gauge has an adjustable stem length from 34.5 to 48 inches and provides high, mid and low level indication. It can handle temperatures up to 250°F and pressures up to 100 PSI. The Krueger Level Gauge is ideal for use with hydraulic fluids, diesel fuel and motor oils.