Learn More
Grainger G3261 Level Gauge HDPE D 99in
Catalog No. 502764021
Encompass_Preferred
The Grainger 16N362 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, 2.5 lb extinguisher uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment and assets. The extinguisher has a discharge range of 10-15 ft and can operate between -4°F to 120°F. It features an all-metal valve construction and comes with a wall hook for easy mounting. This compact, portable extinguisher is well-suited for electronic equipment fires.
Catalog No. 50-276-4021 Supplier Grainger Supplier No. 16N362
The Grainger 16N362 is a 3M Novec 1230 Clean Agent Fire Extinguisher that provides fast fire suppression for Class A, B, and C fires. This UL rated, 2.5 lb extinguisher uses Novec 1230 fluid which leaves no residue and is ideal for protecting valuable equipment and assets. The extinguisher has a discharge range of 10-15 ft and can operate between -4°F to 120°F. It features an all-metal valve construction and comes with a wall hook for easy mounting. This compact, portable extinguisher is well-suited for electronic equipment fires.