Learn More
Grainger G3262 Level Gauge HDPE D 101in
Catalog No. 502764098
Encompass_Preferred
The Fisherbrand G3262 Level Gauge is made from HDPE and has a diameter of 1.03 inches. It features a clear, unbreakable polycarbonate tube with bold black level markings every 1/8 inch. The polypropylene float has a diameter of 0.75 inches and is lightweight. The gauge has 3/8 inch NPT male threaded ends and can withstand temperatures from -20 to 180°F. It has a maximum pressure rating of 150 psi. The gauge measures 16 inches long and weighs 0.25 lbs. This level gauge is ideal for use with water, oil, and dilute acids.
Catalog No. 50-276-4098 Supplier Grainger Supplier No. 16N284
The Fisherbrand G3262 Level Gauge is made from HDPE and has a diameter of 1.03 inches. It features a clear, unbreakable polycarbonate tube with bold black level markings every 1/8 inch. The polypropylene float has a diameter of 0.75 inches and is lightweight. The gauge has 3/8 inch NPT male threaded ends and can withstand temperatures from -20 to 180°F. It has a maximum pressure rating of 150 psi. The gauge measures 16 inches long and weighs 0.25 lbs. This level gauge is ideal for use with water, oil, and dilute acids.