Learn More
Grainger G3262 Level Gauge HDPE D 103in
Catalog No. 502764507
Encompass_Preferred
The Grainger 16N286 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The 16N286 socket adapter is ideal for use in tight spaces where a 3/8-inch drive tool may not fit.
Catalog No. 50-276-4507 Supplier Grainger Supplier No. 16N286
The Grainger 16N286 is a Westward brand 1/4-inch drive socket adapter that converts a 1/4-inch square drive to a 3/8-inch square drive. It is made from chrome vanadium steel for durability and has a black oxide finish to resist corrosion. This socket adapter allows you to use 1/4-inch drive tools with 3/8-inch drive ratchets, handles, extensions, etc. It measures 1-1/2 inches long and weighs 0.8 ounces. The 16N286 socket adapter is ideal for use in tight spaces where a 3/8-inch drive tool may not fit.