Learn More
Grainger G3262 Level Gauge HDPE D 104in
Catalog No. 502764508
Encompass_Preferred
The Krueger Level Gauge is a liquid level indicator designed for use in industrial tanks and vessels. It features a polypropylene float that moves up and down a stainless steel stem in response to liquid level changes. The float's position is indicated by a circular scale and pointer attached to the stem. This mechanical gauge has a maximum measuring range of 60 inches and is suitable for use with water, oil and other non-corrosive liquids. The gauge mounts to the top of the tank through a 3/4 inch NPT connection and requires no external power source. It provides a simple, economical way to monitor liquid levels.
Catalog No. 50-276-4508 Supplier Grainger Supplier No. 16N287
The Krueger Level Gauge is a liquid level indicator designed for use in industrial tanks and vessels. It features a polypropylene float that moves up and down a stainless steel stem in response to liquid level changes. The float's position is indicated by a circular scale and pointer attached to the stem. This mechanical gauge has a maximum measuring range of 60 inches and is suitable for use with water, oil and other non-corrosive liquids. The gauge mounts to the top of the tank through a 3/4 inch NPT connection and requires no external power source. It provides a simple, economical way to monitor liquid levels.