Learn More
Grainger G3262 Level Gauge HDPE D 105in
Catalog No. 502764556
Encompass_Preferred
The Krueger 18-Inch Container Level Gauge is a float-operated level gauge designed for use with 18-inch diameter tanks. It features a hermetically sealed gauge head with a float connected to a stainless steel indicating rod. The rod moves up and down with the liquid level and indicates the level on an external, easy-to-read scale. This heavy duty level gauge is made of corrosion resistant materials and is designed for use with water, chemicals, petroleum products and other liquids. It can handle temperatures up to 250°F. The gauge mounts to the top of the tank and provides continuous monitoring of liquid level.
Catalog No. 50-276-4556 Supplier Grainger Supplier No. 16N288
The Krueger 18-Inch Container Level Gauge is a float-operated level gauge designed for use with 18-inch diameter tanks. It features a hermetically sealed gauge head with a float connected to a stainless steel indicating rod. The rod moves up and down with the liquid level and indicates the level on an external, easy-to-read scale. This heavy duty level gauge is made of corrosion resistant materials and is designed for use with water, chemicals, petroleum products and other liquids. It can handle temperatures up to 250°F. The gauge mounts to the top of the tank and provides continuous monitoring of liquid level.