Learn More
Grainger G3262 Level Gauge HDPE D 106in
Catalog No. 502764610
Encompass_Preferred
The Grainger 16N289 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning, scouring, and finishing. It features an open web construction with non-woven nylon fibers coated in abrasive mineral. This 8" x 4" hand pad has a medium duty abrasiveness making it versatile for various scrubbing and finishing applications. The open web design allows it to conform to surfaces. It can be used wet or dry on surfaces including metal, plastic, glass, and ceramic. The mineral coating cuts through grease and grime without scratching. It is a disposable hand pad.
Catalog No. 50-276-4610 Supplier Grainger Supplier No. 16N289
The Grainger 16N289 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for cleaning, scouring, and finishing. It features an open web construction with non-woven nylon fibers coated in abrasive mineral. This 8" x 4" hand pad has a medium duty abrasiveness making it versatile for various scrubbing and finishing applications. The open web design allows it to conform to surfaces. It can be used wet or dry on surfaces including metal, plastic, glass, and ceramic. The mineral coating cuts through grease and grime without scratching. It is a disposable hand pad.