Learn More
Grainger G3262 Level Gauge HDPE D 107in
Catalog No. 502764624
Encompass_Preferred
The Krueger Level Gauge For 107 in Container is a mechanical level gauge designed for use with 107 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch dial with gallon graduations and a maximum capacity of 550 gallons. This mechanical gauge requires no power and allows visual monitoring of liquid level. It can be mounted on the top or side of compatible containers. The gauge body is 3/4 inch NPT for easy installation.
Catalog No. 50-276-4624 Supplier Grainger Supplier No. 16N290
The Krueger Level Gauge For 107 in Container is a mechanical level gauge designed for use with 107 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge has a 4 inch dial with gallon graduations and a maximum capacity of 550 gallons. This mechanical gauge requires no power and allows visual monitoring of liquid level. It can be mounted on the top or side of compatible containers. The gauge body is 3/4 inch NPT for easy installation.