Learn More
Grainger G3262 Level Gauge HDPE D 109in
Catalog No. 502764126
Encompass_Preferred
The Grainger website features aluminum and HDPE rod/float liquid level gauges for drums and tanks. These gauges allow visual monitoring of liquid levels, with floats moving up and down rods as liquid levels change. They are available in various sizes and configurations to fit different tank openings and applications. Key features include corrosion resistance, chemical compatibility, easy installation, and economical operation. These basic gauges provide a simple, reliable way to monitor liquid levels in drums and tanks.
Catalog No. 50-276-4126 Supplier Grainger Supplier No. 16N291
The Grainger website features aluminum and HDPE rod/float liquid level gauges for drums and tanks. These gauges allow visual monitoring of liquid levels, with floats moving up and down rods as liquid levels change. They are available in various sizes and configurations to fit different tank openings and applications. Key features include corrosion resistance, chemical compatibility, easy installation, and economical operation. These basic gauges provide a simple, reliable way to monitor liquid levels in drums and tanks.