Learn More
Grainger G3262 Level Gauge HDPE D 17in
Catalog No. 502766636
Encompass_Preferred
The Krueger 17-inch Level Gauge is a liquid level indicator designed for 17-inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a useful length of 15 inches and can handle temperatures up to 180°F. It has a maximum pressure rating of 15 PSI. This economical gauge helps monitor liquid levels in a variety of applications.
Catalog No. 50-276-6636 Supplier Grainger Supplier No. 16N213
The Krueger 17-inch Level Gauge is a liquid level indicator designed for 17-inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a useful length of 15 inches and can handle temperatures up to 180°F. It has a maximum pressure rating of 15 PSI. This economical gauge helps monitor liquid levels in a variety of applications.