Learn More
Grainger G3262 Level Gauge HDPE D 18in
Catalog No. 502766605
Encompass_Preferred
The AT-A-GLANCE Level Gauge For 18 in Container is a reusable level indicator that allows you to visually monitor liquid levels in 18 inch diameter tanks or containers. It has a durable polypropylene construction and features large, easy-to-read level markers from FULL to ADD in half inch increments. This device allows operators to quickly check fill levels without opening or unsealing containers. It can withstand temperatures up to 180°F. The AT-A-GLANCE level gauge is an economical solution for monitoring fluid levels in industrial applications.
Catalog No. 50-276-6605 Supplier Grainger Supplier No. 16N214
The AT-A-GLANCE Level Gauge For 18 in Container is a reusable level indicator that allows you to visually monitor liquid levels in 18 inch diameter tanks or containers. It has a durable polypropylene construction and features large, easy-to-read level markers from FULL to ADD in half inch increments. This device allows operators to quickly check fill levels without opening or unsealing containers. It can withstand temperatures up to 180°F. The AT-A-GLANCE level gauge is an economical solution for monitoring fluid levels in industrial applications.