Learn More
Grainger G3262 Level Gauge HDPE D 19in
Catalog No. 502766723
Encompass_Preferred
The Krueger Level Gauge For 19 in Container is a liquid level gauge designed for 19 inch wide containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has color coded zones from empty to full to provide quick visual monitoring of liquid level. This industrial gauge is made in the USA and is compatible with water-based chemicals and petroleum products. The gauge measures 19 inches wide and can be mounted to the container with adhesive.
Catalog No. 50-276-6723 Supplier Grainger Supplier No. 16N215
The Krueger Level Gauge For 19 in Container is a liquid level gauge designed for 19 inch wide containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has color coded zones from empty to full to provide quick visual monitoring of liquid level. This industrial gauge is made in the USA and is compatible with water-based chemicals and petroleum products. The gauge measures 19 inches wide and can be mounted to the container with adhesive.