Learn More
Grainger G3262 Level Gauge HDPE D 23in
Catalog No. 502766791
Encompass_Preferred
The Krueger Level Gauge For 26 in Container is a mechanical level gauge designed for use with 26 inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 22 inches and is suitable for use with water-based chemicals and petroleum products. This mechanical gauge requires no power and allows quick visual monitoring of tank level. With its corrosion resistant construction, this level gauge provides a simple and reliable solution for monitoring liquid levels in 26 inch tanks or containers.
Catalog No. 50-276-6791 Supplier Grainger Supplier No. 16N219
The Krueger Level Gauge For 26 in Container is a mechanical level gauge designed for use with 26 inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 22 inches and is suitable for use with water-based chemicals and petroleum products. This mechanical gauge requires no power and allows quick visual monitoring of tank level. With its corrosion resistant construction, this level gauge provides a simple and reliable solution for monitoring liquid levels in 26 inch tanks or containers.