Learn More
Grainger G3262 Level Gauge HDPE D 26in
Catalog No. 502766705
Encompass_Preferred
The Krueger Level Gauge For 18 in Container is a mechanical level gauge designed for use with 18 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT threads for easy installation and its lightweight design allows it to float right on the liquid surface. This economical gauge is ideal for monitoring liquid levels in a variety of applications.
Catalog No. 50-276-6705 Supplier Grainger Supplier No. 16N221
The Krueger Level Gauge For 18 in Container is a mechanical level gauge designed for use with 18 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT threads for easy installation and its lightweight design allows it to float right on the liquid surface. This economical gauge is ideal for monitoring liquid levels in a variety of applications.