Learn More
Grainger G3262 Level Gauge HDPE D 27in
Catalog No. 502766734
Encompass_Preferred
The At-A-Glance liquid level gauges are designed for use on drums and tanks to indicate liquid level at a glance. They feature a clear polycarbonate body with colored float balls and are available in various sizes to fit standard drum openings. The gauges allow quick visual monitoring of liquid level and can be used with water, oil and various chemicals. They are made in the USA and are durable and reusable. The gauges provide an economical way to monitor liquid levels in drums and tanks across many industries.
Catalog No. 50-276-6734 Supplier Grainger Supplier No. 16N222
The At-A-Glance liquid level gauges are designed for use on drums and tanks to indicate liquid level at a glance. They feature a clear polycarbonate body with colored float balls and are available in various sizes to fit standard drum openings. The gauges allow quick visual monitoring of liquid level and can be used with water, oil and various chemicals. They are made in the USA and are durable and reusable. The gauges provide an economical way to monitor liquid levels in drums and tanks across many industries.