Learn More
Grainger G3262 Level Gauge HDPE D 29in
Catalog No. 502768422
Encompass_Preferred
The Krueger Level Gauge For 29 in Container is a mechanical level gauge designed for use with 29 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/8 inch increments. It has a maximum reading of 27 inches and a minimum reading of 1 3/4 inches. The gauge installs easily by threading into a standard 2 inch opening. It is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-8422 Supplier Grainger Supplier No. 16N224
The Krueger Level Gauge For 29 in Container is a mechanical level gauge designed for use with 29 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/8 inch increments. It has a maximum reading of 27 inches and a minimum reading of 1 3/4 inches. The gauge installs easily by threading into a standard 2 inch opening. It is made in the USA by Krueger Sentry Gauge.