Learn More
Grainger G3262 Level Gauge HDPE D 31in
Catalog No. 502766606
Encompass_Preferred
The Krueger Level Gauge For 133 in Container is a mechanical level gauge designed for use with 133 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates container level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is ideal for visual monitoring of liquid levels in larger containers.
Catalog No. 50-276-6606 Supplier Grainger Supplier No. 16N225
The Krueger Level Gauge For 133 in Container is a mechanical level gauge designed for use with 133 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates container level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT mounting that allows it to be securely installed on the container. This heavy duty mechanical gauge is ideal for visual monitoring of liquid levels in larger containers.