Learn More
Grainger G3262 Level Gauge HDPE D 33in
Supplier:  Grainger 16N227
Encompass_Preferred
The Grainger 33" Liquid Level Gauge for Drums and Tanks is a float-operated mechanical gauge that indicates liquid level for containers up to 33 inches deep. It features a hermetically sealed glass tube with color coded floats to provide an easy visual reference of liquid level. This heavy duty polypropylene gauge has a lightweight aluminum mounting bracket and is designed for use with water, oil and mild chemicals. It can withstand temperatures up to 180°F. The 33" gauge model has a 6" face and mounts to the top of horizontal or vertical tanks.
Catalog No. 50-276-6733