Learn More
Grainger G3262 Level Gauge HDPE D 34in
Catalog No. 502767558
Encompass_Preferred
The Krueger Level Gauge For 34 in Container is a liquid level indicator designed for use with 34 inch high containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is graduated in inches and fractions for easy level reading. This gauge helps monitor liquid levels in containers up to 34 inches tall. It has a maximum temperature rating of 180°F and is made in the USA. The gauge installs by threading into a standard 2 inch opening.
Catalog No. 50-276-7558 Supplier Grainger Supplier No. 16N228
The Krueger Level Gauge For 34 in Container is a liquid level indicator designed for use with 34 inch high containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is graduated in inches and fractions for easy level reading. This gauge helps monitor liquid levels in containers up to 34 inches tall. It has a maximum temperature rating of 180°F and is made in the USA. The gauge installs by threading into a standard 2 inch opening.