Learn More
Grainger G3262 Level Gauge HDPE D 35in
Catalog No. 502765362
Encompass_Preferred
The Krueger Level Gauge For 35 in Container is a mechanical level gauge designed for use with 35 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be screwed into the container. This heavy duty mechanical gauge is ideal for visual monitoring of liquid levels in larger containers.
Catalog No. 50-276-5362 Supplier Grainger Supplier No. 16N229
The Krueger Level Gauge For 35 in Container is a mechanical level gauge designed for use with 35 inch diameter containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked scale in 1/2 inch increments. It has 3/4 inch NPT mounting that allows it to be screwed into the container. This heavy duty mechanical gauge is ideal for visual monitoring of liquid levels in larger containers.