Learn More
Grainger G3262 Level Gauge HDPE D 37in
Catalog No. 502766876
Encompass_Preferred
The Krueger Level Gauge For 37 in Container is a mechanical level gauge designed for 37 inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a dual scale that reads from 0 to 100 percent in 10 percent increments on the front and 0 to 35 inches in 1 inch increments on the back. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge has 3/4 inch NPT connections and is made in the USA.
Catalog No. 50-276-6876 Supplier Grainger Supplier No. 16N230
The Krueger Level Gauge For 37 in Container is a mechanical level gauge designed for 37 inch diameter containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a dual scale that reads from 0 to 100 percent in 10 percent increments on the front and 0 to 35 inches in 1 inch increments on the back. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge has 3/4 inch NPT connections and is made in the USA.