Learn More
Grainger G3262 Level Gauge HDPE D 39in
Catalog No. 502765363
Encompass_Preferred
The Krueger Level Gauge For 39 in Container is a mechanical level gauge designed for use with 39 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 36 inches and is suitable for use with water-based chemicals. This mechanical gauge requires no power and is easy to install by drilling a 1 inch hole in the container wall. It can withstand temperatures up to 180°F. The gauge body measures 43 inches tall.
Catalog No. 50-276-5363 Supplier Grainger Supplier No. 16N231
The Krueger Level Gauge For 39 in Container is a mechanical level gauge designed for use with 39 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 36 inches and is suitable for use with water-based chemicals. This mechanical gauge requires no power and is easy to install by drilling a 1 inch hole in the container wall. It can withstand temperatures up to 180°F. The gauge body measures 43 inches tall.