Learn More
Grainger G3262 Level Gauge HDPE D 47in
Catalog No. 502767094
Encompass_Preferred
The Grainger G3262 Level Gauge is a 47 inch long high density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/8 inch for precise level indication. This dipstick is lightweight, chemical resistant, and can withstand temperatures up to 120°F. With an overall length of 47 inches, this level gauge can measure tank depths up to 46 inches. The HDPE construction ensures durability and chemical compatibility.
Catalog No. 50-276-7094 Supplier Grainger Supplier No. 16N236
The Grainger G3262 Level Gauge is a 47 inch long high density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/8 inch for precise level indication. This dipstick is lightweight, chemical resistant, and can withstand temperatures up to 120°F. With an overall length of 47 inches, this level gauge can measure tank depths up to 46 inches. The HDPE construction ensures durability and chemical compatibility.