Learn More
Grainger G3262 Level Gauge HDPE D 50in
Catalog No. 502767095
Encompass_Preferred
The Krueger Level Gauge For 108 in Container is a liquid level gauge designed for 108 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 108 inches and includes markings every 1/4 inch for precise measurement. This industrial gauge helps monitor liquid levels in large tanks and containers. With its durable construction, it provides reliable liquid level indication for operations.
Catalog No. 50-276-7095 Supplier Grainger Supplier No. 16N238
The Krueger Level Gauge For 108 in Container is a liquid level gauge designed for 108 inch tall containers. It features a polypropylene float that moves up and down a stainless steel tape to indicate liquid level. The gauge has a maximum reading of 108 inches and includes markings every 1/4 inch for precise measurement. This industrial gauge helps monitor liquid levels in large tanks and containers. With its durable construction, it provides reliable liquid level indication for operations.