Learn More
Grainger G3262 Level Gauge HDPE D 56in
Catalog No. 502767110
Encompass_Preferred
The Grainger G3262 Level Gauge is a 56 inch long high-density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/8 inch for precise level indication. This durable HDPE construction stands up to oils, solvents and other chemicals. The tapered end easily guides into fill ports and openings. Overall length is 56 inches and diameter is 1/4 inch. This handy dipstick level gauge provides accurate liquid level readings for a variety of applications.
Catalog No. 50-276-7110 Supplier Grainger Supplier No. 16N244
The Grainger G3262 Level Gauge is a 56 inch long high-density polyethylene dipstick designed for measuring liquid levels in tanks and reservoirs. It features easy-to-read black markings every 1/8 inch for precise level indication. This durable HDPE construction stands up to oils, solvents and other chemicals. The tapered end easily guides into fill ports and openings. Overall length is 56 inches and diameter is 1/4 inch. This handy dipstick level gauge provides accurate liquid level readings for a variety of applications.