Learn More
Grainger G3262 Level Gauge HDPE D 57in
Catalog No. 502766762
Encompass_Preferred
The Krueger Therma Level Gauge For 57-16N606 is a liquid level gauge designed for use with Krueger air handlers. It features a clear polycarbonate gauge body to allow easy viewing of refrigerant level. This gauge has a temperature range of 0 to 250°F, a working pressure of 435 psi, and 1/4" FPT connections. The gauge body measures 2-1/2" in diameter x 5-1/2" long. This item is sold individually.
Catalog No. 50-276-6762 Supplier Grainger Supplier No. 16N245
The Krueger Therma Level Gauge For 57-16N606 is a liquid level gauge designed for use with Krueger air handlers. It features a clear polycarbonate gauge body to allow easy viewing of refrigerant level. This gauge has a temperature range of 0 to 250°F, a working pressure of 435 psi, and 1/4" FPT connections. The gauge body measures 2-1/2" in diameter x 5-1/2" long. This item is sold individually.