Learn More
Grainger G3262 Level Gauge HDPE D 61in
Catalog No. 502766782
Encompass_Preferred
The G3262 Level Gauge HDPE D 61in 16N248 is a 61 inch long high density polyethylene dipstick level gauge made by Grainger. It has a 1/4 inch diameter and is used to measure liquid levels in tanks and reservoirs. The gauge is graduated in inches and centimeters for easy reading. It has a maximum temperature rating of 140°F and is resistant to oils, solvents and other chemicals. The level gauge provides an economical way to monitor liquid levels.
Catalog No. 50-276-6782 Supplier Grainger Supplier No. 16N248
The G3262 Level Gauge HDPE D 61in 16N248 is a 61 inch long high density polyethylene dipstick level gauge made by Grainger. It has a 1/4 inch diameter and is used to measure liquid levels in tanks and reservoirs. The gauge is graduated in inches and centimeters for easy reading. It has a maximum temperature rating of 140°F and is resistant to oils, solvents and other chemicals. The level gauge provides an economical way to monitor liquid levels.