Learn More
Grainger G3262 Level Gauge HDPE D 62in
Catalog No. 502766781
Encompass_Preferred
The Krueger 62-inch Level Gauge is a liquid level indicator designed for 62-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a dual scale that reads from 0 to 62 inches in 1/8 inch increments on one side and 0 to 5 feet in 1/10 foot increments on the other. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge installs easily using a 1-inch NPT connection and is ideal for use with water treatment tanks, chemical tanks, day tanks, storage tanks and more.
Catalog No. 50-276-6781 Supplier Grainger Supplier No. 16N249
The Krueger 62-inch Level Gauge is a liquid level indicator designed for 62-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a dual scale that reads from 0 to 62 inches in 1/8 inch increments on one side and 0 to 5 feet in 1/10 foot increments on the other. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge installs easily using a 1-inch NPT connection and is ideal for use with water treatment tanks, chemical tanks, day tanks, storage tanks and more.