Learn More
Grainger G3262 Level Gauge HDPE D 63in
Catalog No. 502764101
Encompass_Preferred
The Grainger 63" Liquid Level Gauge is designed for use in 63" deep drums and tanks to visually indicate liquid level. It features a polypropylene float and 316 stainless steel guide wire with a max temperature rating of 180°F. The gauge has bold black measurement markings in 1/4" increments for easy viewing. This economical gauge provides accurate monitoring of liquid levels in drums up to 63" deep. It can help prevent overfilling and spills.
Catalog No. 50-276-4101 Supplier Grainger Supplier No. 16N250
Add to Cart
Add to Cart
The Grainger 63" Liquid Level Gauge is designed for use in 63" deep drums and tanks to visually indicate liquid level. It features a polypropylene float and 316 stainless steel guide wire with a max temperature rating of 180°F. The gauge has bold black measurement markings in 1/4" increments for easy viewing. This economical gauge provides accurate monitoring of liquid levels in drums up to 63" deep. It can help prevent overfilling and spills.