Learn More
Grainger G3262 Level Gauge HDPE D 65in
Catalog No. 502764467
Encompass_Preferred
The Grainger G3262 Level Gauge is a 65 inch long dipstick made from durable high density polyethylene plastic. It features easy-to-read black measurement markings every 1/8 inch for precise level indication. This dipstick can be used for measuring fluid levels in tanks, reservoirs, and other containers. It has a maximum temperature rating of 120°F and is chemically resistant to gasoline, oil, and other petroleum products. The G3262 provides an economical way to check fluid levels.
Catalog No. 50-276-4467 Supplier Grainger Supplier No. 16N251
The Grainger G3262 Level Gauge is a 65 inch long dipstick made from durable high density polyethylene plastic. It features easy-to-read black measurement markings every 1/8 inch for precise level indication. This dipstick can be used for measuring fluid levels in tanks, reservoirs, and other containers. It has a maximum temperature rating of 120°F and is chemically resistant to gasoline, oil, and other petroleum products. The G3262 provides an economical way to check fluid levels.