Learn More
Grainger G3262 Level Gauge HDPE D 66in
Catalog No. 502764531
Encompass_Preferred
The Krueger Level Gauge For 66 in Container is a mechanical level gauge designed for use with 66 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 60 inches and includes markings every 5 inches. This heavy duty industrial gauge can withstand temperatures up to 180°F and fits 2 inch bung openings. It is made in the USA by Krueger Sentry Gauge.
Catalog No. 50-276-4531 Supplier Grainger Supplier No. 16N252
Add to Cart
Add to Cart
The Krueger Level Gauge For 66 in Container is a mechanical level gauge designed for use with 66 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 60 inches and includes markings every 5 inches. This heavy duty industrial gauge can withstand temperatures up to 180°F and fits 2 inch bung openings. It is made in the USA by Krueger Sentry Gauge.