Learn More
Grainger G3262 Level Gauge HDPE D 67in
Catalog No. 502763700
Encompass_Preferred
The Grainger 67" Liquid Level Gauge is designed for use in drums and tanks up to 67" deep to visually indicate liquid level. It features a clear polycarbonate body with easy-to-read level markings and a 3/4" NPT male connection. The float is constructed of corrosion resistant 316 stainless steel and it can withstand temperatures up to 200°F. This economical gauge provides accurate level indication for water, oil and other non-corrosive liquids. With proper installation, it allows quick visual monitoring to prevent overfills or run-outs.
Catalog No. 50-276-3700 Supplier Grainger Supplier No. 16N253
The Grainger 67" Liquid Level Gauge is designed for use in drums and tanks up to 67" deep to visually indicate liquid level. It features a clear polycarbonate body with easy-to-read level markings and a 3/4" NPT male connection. The float is constructed of corrosion resistant 316 stainless steel and it can withstand temperatures up to 200°F. This economical gauge provides accurate level indication for water, oil and other non-corrosive liquids. With proper installation, it allows quick visual monitoring to prevent overfills or run-outs.