Learn More
Grainger G3262 Level Gauge HDPE D 69in
Catalog No. 502764494
Encompass_Preferred
The Grainger HDPE Level Gauge is a 69 inch long dipstick made of durable high density polyethylene plastic. It features inch and 1/8 inch graduations for accurate liquid level measurement in tanks up to 60 inches deep. The gauge's flat design allows it to fit through narrow tank openings and the HDPE material is chemical resistant. Overall, this is a heavy duty dipstick level gauge for measuring liquid levels in industrial tanks and containers.
Catalog No. 50-276-4494 Supplier Grainger Supplier No. 16N255
The Grainger HDPE Level Gauge is a 69 inch long dipstick made of durable high density polyethylene plastic. It features inch and 1/8 inch graduations for accurate liquid level measurement in tanks up to 60 inches deep. The gauge's flat design allows it to fit through narrow tank openings and the HDPE material is chemical resistant. Overall, this is a heavy duty dipstick level gauge for measuring liquid levels in industrial tanks and containers.