Learn More
Grainger G3262 Level Gauge HDPE D 71in
Catalog No. 502764455
Encompass_Preferred
The Krueger 71-inch Level Gauge is a liquid level indicator designed for 71-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is marked with gallon increments from 0 to 550 gallons. This gauge can handle temperatures up to 150°F and pressures up to 100 PSI. It has 3/4 inch NPT thread connections. The float has a specific gravity of 0.75. This economical gauge provides accurate liquid level indication for a variety of applications.
Catalog No. 50-276-4455 Supplier Grainger Supplier No. 16N257
The Krueger 71-inch Level Gauge is a liquid level indicator designed for 71-inch containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The tube is marked with gallon increments from 0 to 550 gallons. This gauge can handle temperatures up to 150°F and pressures up to 100 PSI. It has 3/4 inch NPT thread connections. The float has a specific gravity of 0.75. This economical gauge provides accurate liquid level indication for a variety of applications.