Learn More
Grainger G3262 Level Gauge HDPE D 73in
Catalog No. 502763701
Encompass_Preferred
The Krueger 73-inch Level Gauge is a liquid level indicator designed for 73-inch containers. It features a polypropylene float that moves up and down a stainless steel tape to show the liquid level. The gauge has a maximum reading of 73 inches and includes markings every 1/4 inch for precise measurement. The polypropylene float provides corrosion resistance while the stainless steel tape allows for strength and easy cleaning. This industrial gauge is ideal for measuring liquid levels in large tanks and containers up to 73 inches tall. It can handle temperatures up to 180°F. The gauge attaches via mounting clamps sold separately.
Catalog No. 50-276-3701 Supplier Grainger Supplier No. 16N258
The Krueger 73-inch Level Gauge is a liquid level indicator designed for 73-inch containers. It features a polypropylene float that moves up and down a stainless steel tape to show the liquid level. The gauge has a maximum reading of 73 inches and includes markings every 1/4 inch for precise measurement. The polypropylene float provides corrosion resistance while the stainless steel tape allows for strength and easy cleaning. This industrial gauge is ideal for measuring liquid levels in large tanks and containers up to 73 inches tall. It can handle temperatures up to 180°F. The gauge attaches via mounting clamps sold separately.