Learn More
Grainger G3262 Level Gauge HDPE D 75in
Catalog No. 502764051
Encompass_Preferred
The Krueger Level Gauge For 75 in Container is a mechanical level gauge designed for use with 75 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT threads for easy installation and its lightweight design allows it to float on top of liquid. This economical mechanical gauge is ideal for monitoring liquid levels in processing tanks up to 75 inches tall.
Catalog No. 50-276-4051 Supplier Grainger Supplier No. 16N260
The Krueger Level Gauge For 75 in Container is a mechanical level gauge designed for use with 75 inch tall containers. It features a stainless steel body and polypropylene float to provide chemical resistance. The gauge indicates liquid level on a clearly marked external scale in 1/8 inch increments. It has 3/4 inch NPT threads for easy installation and its lightweight design allows it to float on top of liquid. This economical mechanical gauge is ideal for monitoring liquid levels in processing tanks up to 75 inches tall.