Learn More
Grainger G3262 Level Gauge HDPE D 76in
Supplier:  Grainger 16N261
Encompass_Preferred
The Krueger 76-inch Level Gauge is a liquid level indicator designed for 76-inch containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to show the liquid level. The gauge has a dual scale that reads from 0 to 60 inches on one side and 0 to 150cm on the other. It can handle temperatures up to 180°F and pressures up to 100 PSI. The gauge installs easily using a 3/4-inch NPT fitting and requires a 3/4-inch full coupling for installation. It is ideal for use with water treatment tanks, chemical tanks, plating tanks and more.
Catalog No. 50-276-4577