Learn More
Grainger G3262 Level Gauge HDPE D 77in
Catalog No. 502764452
Encompass_Preferred
The Grainger 77" Liquid Level Gauge is designed for use in 77" deep containers to visually indicate liquid level. It features a clear polycarbonate body to provide a wide viewing window and allow users to easily monitor liquid levels. The gauge attaches via a 3/4" NPT connection and is made of chemical resistant materials for compatibility with various liquids. With no moving parts, this model offers a maintenance-free way to monitor tank fill levels. The 77" length is ideal for common 330 gallon IBC totes.
Catalog No. 50-276-4452 Supplier Grainger Supplier No. 16N262
The Grainger 77" Liquid Level Gauge is designed for use in 77" deep containers to visually indicate liquid level. It features a clear polycarbonate body to provide a wide viewing window and allow users to easily monitor liquid levels. The gauge attaches via a 3/4" NPT connection and is made of chemical resistant materials for compatibility with various liquids. With no moving parts, this model offers a maintenance-free way to monitor tank fill levels. The 77" length is ideal for common 330 gallon IBC totes.