Learn More
Grainger G3262 Level Gauge HDPE D 78in
Catalog No. 502764102
Encompass_Preferred
The Krueger Level Gauge For 78 in Container is a mechanical level gauge designed for use with 78 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a scale range from 0 to 78 inches and is suitable for use with water, wastewater, chemicals, petroleum products and food processing liquids. This economical mechanical gauge requires no power and is made in the USA.
Catalog No. 50-276-4102 Supplier Grainger Supplier No. 16N263
The Krueger Level Gauge For 78 in Container is a mechanical level gauge designed for use with 78 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a scale range from 0 to 78 inches and is suitable for use with water, wastewater, chemicals, petroleum products and food processing liquids. This economical mechanical gauge requires no power and is made in the USA.