Learn More
Grainger G3262 Level Gauge HDPE D 79in
Catalog No. 502764578
Encompass_Preferred
The Krueger Level Gauge For 79 in Container is a mechanical level gauge designed for use with 79 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 60 inches and includes markings every 5 inches. This direct reading gauge allows operators to quickly check liquid levels in containers up to 79 inches tall. With its durable stainless steel and polypropylene construction, it provides reliable level indication in a variety of liquids.
Catalog No. 50-276-4578 Supplier Grainger Supplier No. 16N264
The Krueger Level Gauge For 79 in Container is a mechanical level gauge designed for use with 79 inch tall containers. It features a stainless steel body with a polypropylene float that moves up and down a center guide rod to indicate liquid level. The gauge has a maximum reading of 60 inches and includes markings every 5 inches. This direct reading gauge allows operators to quickly check liquid levels in containers up to 79 inches tall. With its durable stainless steel and polypropylene construction, it provides reliable level indication in a variety of liquids.