Learn More
Grainger G3262 Level Gauge HDPE D 81in
Catalog No. 502764592
Encompass_Preferred
The Grainger liquid level gauges for drums and tanks are designed for use in 81" deep containers. They feature polypropylene and stainless steel construction with float balls to indicate liquid level. The gauges have maximum temperature ratings from 200-500°F depending on the specific model. They are available with standard or custom length cables to fit different container depths. The gauges provide at-a-glance monitoring of liquid levels in drums, totes, and other industrial containers. They are an economical solution for basic liquid level indication in chemical, industrial, and other applications.
Catalog No. 50-276-4592 Supplier Grainger Supplier No. 16N266
Add to Cart
Add to Cart
The Grainger liquid level gauges for drums and tanks are designed for use in 81" deep containers. They feature polypropylene and stainless steel construction with float balls to indicate liquid level. The gauges have maximum temperature ratings from 200-500°F depending on the specific model. They are available with standard or custom length cables to fit different container depths. The gauges provide at-a-glance monitoring of liquid levels in drums, totes, and other industrial containers. They are an economical solution for basic liquid level indication in chemical, industrial, and other applications.