Learn More
Grainger G3262 Level Gauge HDPE D 82in
Catalog No. 502764105
Encompass_Preferred
The Grainger liquid level gauges for drums and tanks are designed for use in 82-inch deep containers. They feature polypropylene and stainless steel construction with plastic sight tubes that allow easy monitoring of liquid levels. The gauges have 3/4 inch NPT thread connections and include either brass or stainless steel fittings. They are available in lengths from 34 to 78 inches to accommodate various container depths. The gauges provide accurate, at-a-glance monitoring for applications like chemical storage, waste oil tanks and more.
Catalog No. 50-276-4105 Supplier Grainger Supplier No. 16N267
The Grainger liquid level gauges for drums and tanks are designed for use in 82-inch deep containers. They feature polypropylene and stainless steel construction with plastic sight tubes that allow easy monitoring of liquid levels. The gauges have 3/4 inch NPT thread connections and include either brass or stainless steel fittings. They are available in lengths from 34 to 78 inches to accommodate various container depths. The gauges provide accurate, at-a-glance monitoring for applications like chemical storage, waste oil tanks and more.