Learn More
Grainger G3262 Level Gauge HDPE D 83in
Catalog No. 502763702
Encompass_Preferred
The Krueger Level Gauge For 83 in Container is a liquid level gauge designed for 83 inch tall containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has a maximum liquid temperature rating of 180°F and a maximum specific gravity rating of 1.9. It has a 3/4 inch NPT bottom fitting and includes a vent plug and retaining clip. The overall length is 94 inches. This industrial gauge helps monitor liquid levels in large containers.
Catalog No. 50-276-3702 Supplier Grainger Supplier No. 16N268
The Krueger Level Gauge For 83 in Container is a liquid level gauge designed for 83 inch tall containers. It features a polypropylene float that moves up and down a polycarbonate indicator tube as liquid level changes. The gauge has a maximum liquid temperature rating of 180°F and a maximum specific gravity rating of 1.9. It has a 3/4 inch NPT bottom fitting and includes a vent plug and retaining clip. The overall length is 94 inches. This industrial gauge helps monitor liquid levels in large containers.