Learn More
Grainger G3262 Level Gauge HDPE D 88in
Catalog No. 502764085
Encompass_Preferred
The Grainger G3262 Level Gauge is a durable HDPE dipstick designed for measuring liquid levels in tanks and reservoirs. It features large, easy-to-read markings in both inches and centimeters for level readings up to 88 inches. The gauge's heavy-duty HDPE construction ensures longevity while resisting corrosion, chemicals, and weather. With its float, this portable dipstick allows quick and accurate monitoring of liquid levels in a variety of applications. The G3262 helps identify leaks, facilitates fluid additions and changes, and assists in maintaining optimal fill levels.
Catalog No. 50-276-4085 Supplier Grainger Supplier No. 16N272
The Grainger G3262 Level Gauge is a durable HDPE dipstick designed for measuring liquid levels in tanks and reservoirs. It features large, easy-to-read markings in both inches and centimeters for level readings up to 88 inches. The gauge's heavy-duty HDPE construction ensures longevity while resisting corrosion, chemicals, and weather. With its float, this portable dipstick allows quick and accurate monitoring of liquid levels in a variety of applications. The G3262 helps identify leaks, facilitates fluid additions and changes, and assists in maintaining optimal fill levels.